Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000079.3 | NP_000070.1 | 2 | 176 | ||
NM_001039523.2 | NP_001034612.1 | 2 | 176 | |||
XM_017003256.1 | XP_016858745.1 | 1 | 346 | |||
XM_017003257.1 | XP_016858746.1 | 1 | 343 | |||
GenBank mRNA | AK291338.1 | - | 2 | 117 | ||
AK299445.1 | - | 2 | 165 | |||
AK315312.1 | - | 2 | 176 | |||
BG828551.1 | - | 152 | ||||
CD013889.1 | - | 2 | 116 | |||
DQ323657.1 | - | 1 | 24 | |||
DQ323658.1 | - | 1 | 24 | |||
S77094.1 | - | 2 | 146 | |||
Y00762.1 | - | 2 | 146 |