Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018.4 | NP_001009.1 | 3 | 166 | ||
NM_001308226.1 | NP_001295155.1 | 3 | 257 | |||
GenBank mRNA | BC064908.1 | - | 1 | 110 | 6053159 | |
BC105810.1 | - | 3 | 144 | 6744553 | ||
BC141832.1 | - | 1 | 110 | 40148744 | ||
BE798984.1 | - | 3 | 346 | |||
BG253776.1 | - | 1 | 105 | |||
BG705752.1 | - | 3 | 264 | |||
BM558707.1 | - | 3 | 245 | |||
BU602036.1 | - | 3 | 167 | |||
J02984.1 | - | 3 | 133 |