Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030.4 | NP_001021.1 | 1 | 157 | ||
GenBank mRNA | AK312070.1 | - | 2, 3 | 158 | ||
AV763942.1 | - | 1 | 158 | |||
BC002658.2 | - | 1 | 140 | 3607957 | ||
BC070219.1 | - | 158 | 6744779 | |||
CN304370.1 | - | 1 | 153 | |||
L19739.1 | - | 2, 3 | 143 | |||
U57847.1 | - | 2, 3 | 136 |