Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281.2 | NP_001272.2 | 1, 2 | 691 | ||
GenBank mRNA | AF013488.1 | - | 1, 2 | 181 | ||
BC005969.1 | - | 1, 2 | 168 | 4076810 | ||
BC052812.1 | - | 1, 2 | 191 | 6527446 | ||
BT007424.1 | - | 1, 2 | 116 | |||
D49738.1 | - | 1, 2 | 236 |