Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001540.3 | NP_001531.1 | 3 | 688 | ||
GenBank mRNA | AB020027.1 | - | 3 | 558 | ||
AK296890.1 | - | 4 | 468 | |||
AK311446.1 | - | 3 | 320 | |||
AK311894.1 | - | 3 | 571 | |||
AL050380.1 | - | 4 | 419 | |||
BC000510.2 | - | 3 | 632 | 2822325 | ||
BC012292.1 | - | 3 | 606 | 3862626 | ||
BC012768.2 | - | 3 | 555 | 3689220 | ||
BC014920.1 | - | 3 | 492 | |||
BC073768.1 | - | 3 | 553 | 5444848 | ||
BT019888.1 | - | 3 | 533 | |||
CR407614.1 | - | 3 | 533 | |||
CR536489.1 | - | 3 | 533 | |||
FJ224323.1 | - | 3 | 640 | |||
S74571.1 | - | 2 | 170 | |||
U90906.1 | - | 3 | 640 | |||
X16477.1 | - | 3 | 209 | |||
X54079.1 | - | 3 | 585 |