Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199199.1 | NP_001186128.1 | 2 | 192 | ||
NM_001319139.1 | NP_001306068.1 | 3 | 310 | |||
NM_001319140.1 | NP_001306069.1 | 3 | 310 | |||
NM_001319141.1 | NP_001306070.1 | 2 | 192 | |||
NM_001319143.1 | NP_001306072.1 | 2 | 192 | |||
NM_001625.3 | NP_001616.1 | 2 | 192 | |||
NM_013411.4 | NP_037543.1 | 2 | 192 | |||
NR_134976.1 | 2 | 192 | ||||
GenBank mRNA | AB005621.1 | - | 2 | 147 | ||
AB005622.1 | - | 2 | 151 | |||
AB451267.1 | - | 2 | 109 | |||
AB451394.1 | - | 2 | 109 | |||
AK291676.1 | - | 2 | 129 | |||
AK295105.1 | - | 3 | 243 | |||
AK296863.1 | - | 2 | 126 | |||
AK297036.1 | - | 2 | 129 | |||
AL557311.3 | - | 2 | 166 | |||
AY080899.1 | - | 2 | 144 | |||
AY080900.1 | - | 3 | 230 | |||
BC009405.2 | - | 2 | 120 | 4304794 | ||
BC026705.1 | - | 2 | 128 | |||
BC070127.1 | - | 2 | 134 | 6663039 | ||
BC090040.1 | - | 2 | 134 | 6471696 | ||
BQ889289.1 | - | 3 | 276 | |||
BX504559.1 | - | 2 | 173 | |||
DC422465.1 | - | 2 | 192 | |||
DN601721.1 | - | 2 | 111 | |||
HY061267.1 | - | 2 | 193 | |||
U39945.1 | - | 2 | 142 | |||
U54645.1 | - | 2 | 112 | |||
U84371.1 | - | 2 | 140 |