Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291345.1 | NP_001278274.1 | 5 | 551 | ||
NM_001787.2 | NP_001778.2 | 5 | 581 | |||
NM_033486.2 | NP_277021.2 | 5 | 581 | |||
NM_033487.2 | NP_277022.1 | 5 | 581 | |||
NM_033489.2 | NP_277024.2 | 6 | 628 | |||
NM_033490.2 | NP_277025.1 | 5 | 581 | |||
XM_006711065.3 | XP_006711128.1 | 5 | 1293 | |||
XM_011542490.2 | XP_011540792.1 | 5 | 1323 | |||
XM_011542491.2 | XP_011540793.1 | 5 | 779 | |||
XM_011542492.2 | XP_011540794.1 | 5 | 630 | |||
XM_011542493.2 | XP_011540795.1 | 5 | 1293 | |||
XM_011542494.2 | XP_011540796.1 | 6 | 1370 | |||
XM_011542495.2 | XP_011540797.1 | 6 | 1340 | |||
XM_017002925.1 | XP_016858414.1 | 5 | 1323 | |||
XM_017002926.1 | XP_016858415.1 | 5 | 1293 | |||
XM_017002927.1 | XP_016858416.1 | 5 | 1246 | |||
XM_017002928.1 | XP_016858417.1 | 6 | 1370 | |||
GenBank mRNA | AF067512.1 | - | 5 | 495 | ||
AF067513.1 | - | 5 | 495 | |||
AF067514.1 | - | 6 | 509 | |||
AF067515.1 | - | 5 | 495 | |||
AF067516.1 | - | 6 | 542 | |||
AF067517.1 | - | 5 | 495 | |||
AK000081.1 | - | 4 | 340 | |||
AK295607.1 | - | 6 | 503 | |||
AK304679.1 | - | 5 | 459 | |||
BC009375.2 | - | 5 | 492 | |||
BC140714.1 | - | 5 | 454 | 9021582 | ||
BC171773.1 | - | 5 | 454 | 9054427 | ||
DC295949.1 | - | 5 | 455 |