Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202233.1 | NP_001189162.1 | 3 | 325 | ||
NM_001202234.1 | NP_001189163.1 | 3 | 1252 | |||
NM_002135.4 | NP_002126.2 | 3 | 414 | |||
NM_173157.2 | NP_775180.1 | 2 | 264 | |||
XM_005268822.3 | XP_005268879.1 | 3 | 394 | |||
XM_005268824.3 | XP_005268881.1 | 2 | 584 | |||
XM_006719363.1 | XP_006719426.1 | 3 | 320 | |||
XM_006719364.3 | XP_006719427.1 | 4 | 391 | |||
XM_017019247.1 | XP_016874736.1 | 2 | 196 | |||
GenBank mRNA | AB307717.1 | - | 1 | 95 | ||
AK075054.1 | - | 2 | 223 | |||
AK075491.1 | - | 2 | 114 | |||
AK131566.1 | - | 3 | 1254 | |||
AK297526.1 | - | 3 | 326 | |||
AK314437.1 | - | 2 | 218 | |||
AL702926.1 | - | 2 | 306 | |||
AL831844.1 | - | 1 | 1003 | |||
BC016147.1 | - | 3 | 359 | 3921259 | ||
BT007144.1 | - | 1 | 95 | |||
CR456704.1 | - | 1 | 95 | |||
D49728.1 | - | 2 | 204 | |||
D85245.1 | - | 2 | 285 | |||
HQ692855.1 | - | 1 | 95 | |||
L13740.1 | - | 2 | 205 |