Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002493.4 | NP_002484.1 | 2 | 335 | ||
NM_182739.2 | NP_877416.1 | 2 | 335 | |||
GenBank mRNA | AF035840.1 | - | 2 | 279 | ||
AF067167.1 | - | 2 | 308 | |||
AK289702.1 | - | 2 | 289 | |||
BC009801.1 | - | 2 | 314 | 4334414 | ||
BC016708.1 | - | 2 | 349 | |||
BC029247.1 | - | 2 | 280 | 5171627 | ||
BI669356.1 | - | 2 | 314 | |||
BP208846.1 | - | 2 | 333 | |||
CR456920.1 | - | 2 | 211 |