Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204519.1 | NP_001191448.1 | 1 | 482 | ||
NM_001204520.1 | NP_001191449.1 | 1 | 482 | |||
NM_002561.3 | NP_002552.2 | 1 | 482 | |||
NM_175080.2 | NP_778255.1 | 1 | 482 | |||
GenBank mRNA | AF016709.1 | - | 1 | 119 | ||
AF070573.1 | - | 1 | 110 | |||
AK290889.1 | - | 1 | 482 | |||
AK307959.1 | - | 1 | 313 | |||
BC039015.1 | - | 1 | 223 | 5773706 | ||
BM477082.1 | - | 368 | ||||
DQ234349.1 | - | 1 | 83 | |||
U49395.1 | - | 1 | 135 | |||
U49396.1 | - | 1 | 135 |
Gene Symbol: P2RX5-TAX1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037928.1 | 1 | 482 | |||
GenBank mRNA | AB209623.1 | - | 1 | 272 |