Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000383.3 | NP_000374.1 | 7, 8 | 1059 | ||
GenBank mRNA | AB006682.1 | - | 7, 8 | 1017 | ||
AB006683.1 | - | 2 | 535 | |||
AB006685.1 | - | 2 | 535 | |||
BC137268.1 | - | 1 | 716 | 9021265 | ||
BC137270.1 | - | 1 | 715 | 9021267 | ||
Z97990.1 | - | 7, 8 | 1010 |