Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000554.4 | NP_000545.1 | 3 | 425 | ||
GenBank mRNA | AL711350.1 | - | 3 | 425 | ||
BC016664.1 | - | 3 | 360 | 3869862 | ||
BC053672.1 | - | 3 | 311 | 6144191 | ||
BT007364.1 | - | 2 | 221 |