Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBXA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001060.5 | NP_001051.1 | 3 | 1259 | ||
NM_201636.2 | NP_963998.2 | 3 | 1259 | |||
XM_011528214.2 | XP_011526516.1 | 4 | 3293 | |||
GenBank mRNA | AY429110.1 | - | 2 | 872 | ||
BC074749.2 | - | 2 | 911 | 30915459 | ||
BC074750.2 | - | 2 | 911 | 30915209 | ||
BC121148.1 | - | 2 | 517 | 40121265 | ||
U11271.1 | - | 3 | 1084 | |||
U27325.1 | - | 3 | 995 |
Set Membership: |
G-protein coupled receptor |