Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC25C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287582.1 | NP_001274511.1 | 8, 9 | 1087 | ||
NM_001287583.1 | NP_001274512.1 | 8, 9 | 1204 | |||
NM_001318098.1 | NP_001305027.1 | 8, 9 | 1208 | |||
NM_001790.4 | NP_001781.2 | 8, 9 | 1091 | |||
NM_022809.3 | NP_073720.1 | 5, 6 | 872 | |||
XM_006714739.3 | XP_006714802.1 | 7, 8 | 802 | |||
XM_011543759.2 | XP_011542061.1 | 9, 10 | 875 | |||
XM_011543761.2 | XP_011542063.1 | 5, 6 | 985 | |||
XM_011543762.1 | XP_011542064.1 | 5, 6 | 785 | |||
XM_011543763.1 | XP_011542065.1 | 6, 7 | 500 | |||
XM_011543764.1 | XP_011542066.1 | 5, 6 | 553 | |||
GenBank mRNA | AJ304504.1 | - | 4, 5 | 551 | ||
AK097710.1 | - | 8, 9 | 1204 | |||
AK301828.1 | - | 9, 10 | 1154 | |||
BC019089.2 | - | 8, 9 | 981 | 4858949 | ||
M34065.1 | - | 8, 9 | 980 |