Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199199.1 | NP_001186128.1 | 2 | 266 | ||
NM_001319139.1 | NP_001306068.1 | 3 | 384 | |||
NM_001319140.1 | NP_001306069.1 | 3 | 384 | |||
NM_001319141.1 | NP_001306070.1 | 2 | 266 | |||
NM_001319143.1 | NP_001306072.1 | 2 | 266 | |||
NM_001625.3 | NP_001616.1 | 2 | 266 | |||
NM_013411.4 | NP_037543.1 | 2 | 266 | |||
NR_134976.1 | 2 | 266 | ||||
GenBank mRNA | AB005621.1 | - | 2 | 221 | ||
AB005622.1 | - | 2 | 225 | |||
AB451267.1 | - | 2 | 183 | |||
AB451394.1 | - | 2 | 183 | |||
AK291676.1 | - | 2 | 203 | |||
AK295105.1 | - | 3 | 317 | |||
AK296863.1 | - | 2 | 200 | |||
AK297036.1 | - | 2 | 203 | |||
AL557311.3 | - | 2 | 240 | |||
AY080899.1 | - | 2 | 218 | |||
AY080900.1 | - | 3 | 304 | |||
BC009405.2 | - | 2 | 194 | 4304794 | ||
BC026705.1 | - | 2 | 202 | |||
BC070127.1 | - | 2 | 208 | 6663039 | ||
BC090040.1 | - | 2 | 208 | 6471696 | ||
BQ889289.1 | - | 3 | 350 | |||
BX504559.1 | - | 2 | 247 | |||
DC422465.1 | - | 2 | 266 | |||
DN601721.1 | - | 2 | 185 | |||
HY061267.1 | - | 2 | 267 | |||
U39945.1 | - | 2 | 216 | |||
U54645.1 | - | 2 | 186 | |||
U84371.1 | - | 2 | 214 |