Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172713.1 | NP_001166184.1 | 2 | 478 | ||
NM_002078.4 | NP_002069.2 | 2 | 478 | |||
XM_005265069.3 | XP_005265126.1 | 2 | 398 | |||
XM_005265070.3 | XP_005265127.1 | 2 | 398 | |||
XM_005265071.3 | XP_005265128.1 | 2 | 398 | |||
XM_005265072.3 | XP_005265129.1 | 2 | 398 | |||
XM_005265073.1 | XP_005265130.1 | 2 | 398 | |||
XM_005265074.1 | XP_005265131.1 | 2 | 398 | |||
XM_005265075.1 | XP_005265132.1 | 2 | 400 | |||
XM_006713110.2 | XP_006713173.1 | 2 | 398 | |||
XM_017006184.1 | XP_016861673.1 | 2 | 398 | |||
XM_017006185.1 | XP_016861674.1 | 2 | 888 | |||
XM_017006186.1 | XP_016861675.1 | 2 | 67 | |||
XM_017006187.1 | XP_016861676.1 | 2 | 398 | |||
XM_017006188.1 | XP_016861677.1 | 2 | 400 | |||
GenBank mRNA | AB209693.1 | - | 2 | 393 | ||
AK290550.1 | - | 2 | 400 | |||
AK300192.1 | - | 2 | 398 | |||
AK307429.1 | - | 2 | 398 | |||
BC045543.1 | - | 4 | 1504 | |||
BC050444.1 | - | 2 | 422 | |||
U41740.1 | - | 2 | 403 | |||
X82834.1 | - | 2 | 325 |