Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000157.3 | NP_000148.2 | 8 | 1192 | ||
NM_001005741.2 | NP_001005741.1 | 9 | 1451 | |||
NM_001005742.2 | NP_001005742.1 | 9 | 1432 | |||
NM_001171811.1 | NP_001165282.1 | 7 | 1268 | |||
NM_001171812.1 | NP_001165283.1 | 7 | 1042 | |||
GenBank mRNA | AK291911.1 | - | 9 | 1430 | ||
AK298900.1 | - | 7 | 1042 | |||
AK300186.1 | - | 7 | 1112 | |||
AK300829.1 | - | 7 | 1105 | |||
AK300876.1 | - | 9 | 1432 | |||
AK301374.1 | - | 6 | 801 | |||
AK301879.1 | - | 7 | 771 | |||
AK302000.1 | - | 7 | 1017 | |||
AK312502.1 | - | 9 | 1248 | |||
BC003356.1 | - | 8 | 1131 | 2899915 | ||
BX648487.1 | - | 9 | 1260 | |||
D13286.1 | - | 8 | 1131 | |||
K02920.1 | - | 7 | 1011 | |||
KJ690771.1 | - | 8 | 1075 | |||
KJ690773.1 | - | 8 | 1075 | |||
M16328.1 | - | 8 | 1461 | |||
M19285.1 | - | 8 | 1148 |