Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000905.3 | NP_000896.1 | 3 | 335 | ||
GenBank mRNA | AK312156.1 | - | 3 | 334 | ||
BC029497.1 | - | 3 | 335 | 5278692 | ||
K01911.1 | - | 3 | 333 | |||
M15789.1 | - | 2 | 261 |