Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSTR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001052.2 | NP_001043.2 | 1 | 700 | ||
GenBank mRNA | BC069063.1 | - | 1 | 636 | 7216876 | |
BC117270.1 | - | 1 | 700 | 40125821 | ||
BC117272.1 | - | 1 | 700 | 40125823 |
Set Membership: |
G-protein coupled receptor |