Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF016492.1 | - | 6 | 1638 | ||
BC069441.1 | - | 6 | 1638 | 7262335 | ||
BC107059.2 | - | 6 | 1638 | 40006084 | ||
BC107060.2 | - | 6 | 1638 | 40006085 |