Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002260.3 | NP_002251.2 | 6 | 734 | ||
GenBank mRNA | BC106069.1 | - | 6 | 686 | ||
X54869.1 | - | 6 | 734 |