Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX2-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002509.3 | NP_002500.1 | 2 | 1616 | ||
XM_006723566.3 | XP_006723629.1 | 2 | 1273 |