Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003214.3 | NP_003205.2 | 2 | 415 | ||
GenBank mRNA | AF142482.1 | - | 2 | 369 | ||
BC010053.2 | - | 2 | 271 | |||
BC027877.1 | - | 2 | 281 | 5207501 | ||
BC091488.1 | - | 2 | 340 | 6187474 | ||
BQ719548.1 | - | 2 | 339 | |||
DA856821.1 | - | 2 | 438 | |||
X94439.1 | - | 1 | 179 |