Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003404.4 | NP_003395.1 | 3 | 591 | ||
NM_139323.3 | NP_647539.1 | 2 | 496 | |||
XM_017028039.1 | XP_016883528.1 | 3 | 395 | |||
XM_017028040.1 | XP_016883529.1 | 2 | 389 | |||
GenBank mRNA | AB209759.1 | - | 2 | 2237 | ||
AB451260.1 | - | 1 | 205 | |||
AB451386.1 | - | 1 | 205 | |||
AF064847.1 | - | 3 | 515 | |||
AK292717.1 | - | 3 | 484 | |||
BC001359.2 | - | 2 | 386 | 3051079 | ||
BF316820.1 | - | 3 | 486 | |||
EU794580.1 | - | 3 | 487 | |||
X57346.1 | - | 3 | 577 |