Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDNRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166055.1 | NP_001159527.1 | 6 | 2511 | ||
NM_001256283.1 | NP_001243212.1 | 7 | 2348 | |||
NM_001957.3 | NP_001948.1 | 8 | 2838 | |||
NR_045958.1 | 7 | 2639 | ||||
GenBank mRNA | BC022511.1 | - | 8 | 2659 | 4812050 | |
D90348.1 | - | 8 | 2787 | |||
S57498.1 | - | 8 | 2791 | |||
X61950.1 | - | 8 | 2791 |