Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFITM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003641.3 | NP_003632.3 | 2 | 519 | ||
GenBank mRNA | AK290480.1 | - | 2 | 494 | ||
BC000897.1 | - | 2 | 469 | 3464598 | ||
BT007173.1 | - | 2 | 339 | |||
J04164.1 | - | 2 | 655 | |||
X84958.1 | - | 2 | 449 |