Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003806.2 | NP_003797.1 | 1, 2 | 454 | ||
GenBank mRNA | D83699.1 | - | 1, 2 | 59 | ||
U76376.1 | - | 1, 2 | 454 |