Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004160.5 | NP_004151.3 | 5, 6 | 735 | ||
XM_011525035.1 | XP_011523337.1 | 5, 6 | 1345 | |||
GenBank mRNA | BC041057.1 | - | 5, 6 | 735 | 5441270 | |
CR542129.1 | - | 1, 2 | 194 | |||
D13899.1 | - | 2, 3 | 274 | |||
D13902.1 | - | 2, 3 | 276 |