Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032296.3 | NP_001027467.2 | 2 | 469 | ||
NM_001286649.1 | NP_001273578.1 | 2 | 469 | |||
NM_003576.4 | NP_003567.2 | 2 | 413 | |||
XM_005254079.4 | XP_005254136.1 | 2 | 290 | |||
XM_017020794.1 | XP_016876283.1 | 2 | 586 | |||
XM_017020795.1 | XP_016876284.1 | 2 | 248 | |||
GenBank mRNA | AF024636.1 | - | 2 | 182 | ||
AF083420.1 | - | 2 | 285 | |||
AK297583.1 | - | 2 | 185 | |||
AK299140.1 | - | 2 | 186 | |||
BC035578.1 | - | 2 | 417 | 5539375 | ||
BC065378.1 | - | 2 | 413 | 6598075 | ||
BT019619.1 | - | 2 | 104 | |||
CB996892.1 | - | 496 | ||||
CR542227.1 | - | 2 | 104 | |||
DA386652.1 | - | 2 | 375 | |||
DA395717.1 | - | 2 | 413 | |||
EU668350.1 | - | 2 | 460 |