Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002801.3 | NP_002792.1 | 4 | 439 | ||
GenBank mRNA | AK225658.1 | - | 4 | 412 | ||
AK298012.1 | - | 2 | 747 | |||
AK312208.1 | - | 4 | 399 | |||
BC052369.1 | - | 4 | 377 | 4054165 | ||
BP289065.1 | - | 4 | 439 | |||
BT019723.1 | - | 4 | 311 | |||
Y13640.1 | - | 4 | 399 |