Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003009.2 | NP_003000.1 | 4 | 357 | ||
GenBank mRNA | AF015283.1 | - | 4 | 206 | ||
AK026717.1 | - | 3 | 1919 | |||
BC000581.2 | - | 4 | 222 | 3162433 | ||
BC032546.1 | - | 4 | 247 | 5535970 | ||
BC039597.1 | - | 4 | 225 | 6015153 | ||
BC047893.1 | - | 4 | 224 | 6204763 | ||
BF973456.1 | - | 4 | 370 | |||
U67171.1 | - | 4 | 217 |