Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205206.1 | NP_001192135.1 | 3 | 782 | ||
NM_001205207.1 | NP_001192136.1 | 2 | 531 | |||
NM_194255.2 | NP_919231.1 | 3 | 782 | |||
XM_005261164.3 | XP_005261221.1 | 3 | 755 | |||
XM_011529696.2 | XP_011527998.1 | 4 | 1214 | |||
XM_011529698.2 | XP_011528000.1 | 3 | 818 | |||
XM_011529700.2 | XP_011528002.1 | 3 | 1077 | |||
XM_011529701.2 | XP_011528003.1 | 4 | 1027 | |||
XM_011529702.2 | XP_011528004.1 | 3 | 741 | |||
XM_011529703.2 | XP_011528005.1 | 3 | 809 | |||
XM_011529704.2 | XP_011528006.1 | 3 | 725 | |||
XM_011529706.2 | XP_011528008.1 | 2 | 1170 | |||
XM_011529709.2 | XP_011528011.1 | 3 | 996 | |||
XM_017028443.1 | XP_016883932.1 | 3 | 906 | |||
XM_017028444.1 | XP_016883933.1 | 4 | 1220 | |||
XM_017028445.1 | XP_016883934.1 | 4 | 1790 | |||
XM_017028446.1 | XP_016883935.1 | 3 | 1080 | |||
GenBank mRNA | AF004354.1 | - | 2 | 700 | ||
AK303168.1 | - | 2 | 518 | |||
AK307623.1 | - | 3 | 786 | |||
AK310797.1 | - | 3 | 950 | |||
AK313125.1 | - | 3 | 746 | |||
BC003068.2 | - | 3 | 727 | 3509422 | ||
DC311922.1 | - | 2 | 531 | |||
S78996.1 | - | 2 | 629 | |||
U15939.1 | - | 3 | 723 | |||
U17566.1 | - | 3 | 723 | |||
U19720.1 | - | 3 | 724 |