Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003214.3 | NP_003205.2 | 2, 3 | 439 | ||
GenBank mRNA | AF142482.1 | - | 2, 3 | 393 | ||
BC010053.2 | - | 2, 3 | 295 | |||
BC091488.1 | - | 2, 3 | 364 | 6187474 | ||
BQ719548.1 | - | 2, 3 | 363 | |||
DA856821.1 | - | 2, 3 | 462 | |||
X94439.1 | - | 1, 2 | 203 |