Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195100.1 | NP_001182029.1 | 2 | 706 | ||
NM_080391.3 | NP_536316.1 | 3 | 1107 | |||
XM_005271231.3 | XP_005271288.1 | 3 | 809 | |||
GenBank mRNA | AF208850.1 | - | 2 | 454 | ||
AK292703.1 | - | 2 | 215 | |||
BC018662.1 | - | 3 | 1078 | |||
BC070182.1 | - | 2 | 441 | 4686913 | ||
L39000.1 | - | 2 | 76 | |||
U14603.1 | - | 2 | 536 | |||
U48297.1 | - | 2 | 178 |