Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015881.1 | NP_001015881.1 | 3 | 308 | ||
NM_001318468.1 | NP_001305397.1 | 4 | 776 | |||
NM_001318470.1 | NP_001305399.1 | 4 | 762 | |||
NM_004089.3 | NP_004080.2 | 3 | 513 | |||
NM_198057.2 | NP_932174.1 | 3 | 829 | |||
XM_005262099.1 | XP_005262156.1 | 5 | 790 | |||
XM_005262100.1 | XP_005262157.1 | 4 | 665 | |||
XM_005262102.1 | XP_005262159.1 | 4 | 658 | |||
XM_005262103.3 | XP_005262160.1 | 4 | 768 | |||
XM_011530884.1 | XP_011529186.1 | 4 | 864 | |||
XM_017029335.1 | XP_016884824.1 | 4 | 600 | |||
GenBank mRNA | AB025432.1 | - | 3 | 496 | ||
AF153603.1 | - | 3 | 491 | |||
AF183393.1 | - | 3 | 499 | |||
AF228339.1 | - | 3 | 503 | |||
AK092645.1 | - | 4 | 715 | |||
AK092669.1 | - | 3 | 801 | |||
AK127938.1 | - | 3 | 453 | |||
AK300535.1 | - | 4 | 640 | |||
AL110191.1 | - | 3 | 512 | |||
AY007119.1 | - | 3 | 412 | |||
BC018148.1 | - | 3 | 195 | |||
BC072446.1 | - | 3 | 507 | 6165765 | ||
BQ919216.1 | - | 308 | ||||
BX647854.1 | - | 4 | 762 | |||
CR533450.1 | - | 3 | 251 | |||
CR933650.1 | - | 3 | 515 | |||
Z50781.1 | - | 3 | 227 |