Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004316.3 | NP_004307.2 | 1 | 1035 | ||
GenBank mRNA | AK290539.1 | - | 1 | 1036 | ||
AK314114.1 | - | 1 | 651 | |||
BC001638.2 | - | 1 | 1023 | |||
BC002341.2 | - | 1 | 1017 | 2821173 | ||
BC003134.1 | - | 1 | 1017 | 2821173 | ||
BC004425.1 | - | 1 | 1023 | 2819879 | ||
BC031299.1 | - | 1 | 1001 | 3534115 | ||
L08424.1 | - | 1 | 902 |