Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002027.1 | NP_001002027.1 | 3, 4 | 189 | ||
NM_005175.2 | NP_005166.1 | 3, 4 | 259 | |||
GenBank mRNA | AK311848.1 | - | 3, 4 | 158 | ||
AL080089.1 | - | 3, 4 | 179 | |||
BC004963.2 | - | 2, 3 | 135 | 3536694 | ||
BF573360.1 | - | 3, 4 | 189 | |||
BG611396.1 | - | 169 | ||||
BT007230.1 | - | 2, 3 | 125 | |||
BU596803.1 | - | 210 | ||||
CF129854.1 | - | 3, 4 | 193 | |||
CR533449.1 | - | 2, 3 | 125 | |||
D13118.1 | - | 3, 4 | 169 | |||
M16453.1 | - | 18 |