Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HOXB3 SIRNA Assays ›
Gene Symbol:
HOXB3
Gene Name:
homeobox B3
Gene Aliases:
HOX2, HOX2G, Hox-2.7
Chromosome Location:
Chr.17: 48548870 - 48590369 on Build GRCh38
UniGene ID:
Hs.654560
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4202
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HOXB3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002146.4
NP_002137.4
4
1481
XM_005257277.3
XP_005257334.1
5
1195
XM_005257280.3
XP_005257337.1
5
1106
XM_005257282.3
XP_005257339.1
5
671
XM_006721854.2
XP_006721917.1
5
1213
XM_011524707.2
XP_011523009.1
5
11021
XM_011524708.2
XP_011523010.1
4
10601
XM_011524710.1
XP_011523012.1
5
3421
XM_011524719.1
XP_011523021.1
4
5902
XM_011524720.2
XP_011523022.1
4
8593
XM_011524721.2
XP_011523023.1
4
3725
XM_011524726.2
XP_011523028.1
4
679
XM_017024557.1
XP_016880046.1
5
10769
XM_017024558.1
XP_016880047.1
5
10643
XM_017024559.1
XP_016880048.1
5
10485
XM_017024560.1
XP_016880049.1
5
10729
XM_017024561.1
XP_016880050.1
5
14566
XM_017024562.1
XP_016880051.1
5
10507
XM_017024563.1
XP_016880052.1
6
13162
GenBank mRNA
AK027609.1
-
4
1519
AK291182.1
-
5
1192
AK299226.1
-
5
539
AK299265.1
-
5
938
AK309721.1
-
2
359
AK315102.1
-
3
747
AK316245.1
-
4
714
U59298.1
-
2
2072
X16667.1
-
4
986
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
hematopoietic progenitor cell differentiation
transcription, DNA-templated
anterior/posterior pattern specification
rhombomere development
glossopharyngeal nerve morphogenesis
thyroid gland development
embryonic skeletal system morphogenesis
regulation of neurogenesis
cartilage development
definitive hemopoiesis
face development
RNA polymerase II core promoter sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-cp9cp:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline