Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002918.4 | NP_002909.4 | 21 | 4115 | ||
XM_011528165.2 | XP_011526467.1 | 22 | 4445 | |||
XM_011528166.2 | XP_011526468.1 | 22 | 4381 | |||
XM_011528167.2 | XP_011526469.1 | 22 | 4990 | |||
XM_011528168.2 | XP_011526470.1 | 22 | 4151 | |||
XM_011528169.2 | XP_011526471.1 | 21 | 4041 | |||
XM_011528170.2 | XP_011526472.1 | 21 | 4619 | |||
GenBank mRNA | BC049826.1 | - | 21 | 4076 | 6043247 |