Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205206.1 | NP_001192135.1 | 5 | 1394 | ||
NM_001205207.1 | NP_001192136.1 | 4 | 1143 | |||
NM_194255.2 | NP_919231.1 | 5 | 1394 | |||
XM_005261164.3 | XP_005261221.1 | 5 | 1367 | |||
XM_011529696.2 | XP_011527998.1 | 6 | 1826 | |||
XM_011529698.2 | XP_011528000.1 | 5 | 1430 | |||
XM_011529700.2 | XP_011528002.1 | 5 | 1689 | |||
XM_011529701.2 | XP_011528003.1 | 6 | 1639 | |||
XM_011529702.2 | XP_011528004.1 | 5 | 1353 | |||
XM_011529703.2 | XP_011528005.1 | 5 | 1421 | |||
XM_011529704.2 | XP_011528006.1 | 5 | 1337 | |||
XM_011529706.2 | XP_011528008.1 | 4 | 1782 | |||
XM_011529709.2 | XP_011528011.1 | 5 | 1608 | |||
XM_017028443.1 | XP_016883932.1 | 5 | 1518 | |||
XM_017028444.1 | XP_016883933.1 | 6 | 1832 | |||
XM_017028445.1 | XP_016883934.1 | 6 | 2402 | |||
XM_017028446.1 | XP_016883935.1 | 5 | 1692 | |||
GenBank mRNA | AB209069.1 | - | 3 | 445 | ||
AF004354.1 | - | 4 | 1312 | |||
AK303168.1 | - | 4 | 1130 | |||
AK307623.1 | - | 5 | 1398 | |||
AK310797.1 | - | 5 | 1562 | |||
AK313125.1 | - | 5 | 1358 | |||
BC003068.2 | - | 5 | 1339 | 3509422 | ||
S78996.1 | - | 4 | 1241 | |||
U15939.1 | - | 5 | 1335 | |||
U17566.1 | - | 5 | 1335 | |||
U19720.1 | - | 5 | 1336 |