Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318949.1 | NP_001305878.1 | 9 | 2009 | ||
NM_001318950.1 | NP_001305879.1 | 9 | 2154 | |||
NM_001318951.1 | NP_001305880.1 | 8 | 1936 | |||
NM_003459.4 | NP_003450.2 | 8 | 2072 | |||
XM_005264547.1 | XP_005264604.1 | 7 | 1914 | |||
XM_005264548.3 | XP_005264605.1 | 8 | 1900 | |||
XM_006712100.2 | XP_006712163.1 | 7 | 3260 | |||
XM_011533102.2 | XP_011531404.1 | 8 | 2069 | |||
XM_011533103.2 | XP_011531405.1 | 7 | 1929 | |||
XM_017004873.1 | XP_016860362.1 | 7 | 1795 | |||
XM_017004874.1 | XP_016860363.1 | 8 | 2043 | |||
XM_017004875.1 | XP_016860364.1 | 8 | 1827 | |||
GenBank mRNA | AK127096.1 | - | 3 | 2703 | ||
AK127300.1 | - | 4 | 2541 | |||
BC028358.2 | - | 8 | 2060 | 4837595 |