Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004126.3 | NP_004117.1 | 1, 2 | 539 | ||
GenBank mRNA | AF493878.1 | - | 1, 2 | 105 | ||
AK312037.1 | - | 1, 2 | 189 | |||
BC009709.1 | - | 1, 2 | 456 | 3879341 | ||
BT007141.1 | - | 1, 2 | 105 | |||
U31384.1 | - | 1, 2 | 212 |