Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004981.1 | NP_004972.1 | 2 | 1095 | ||
NM_152868.2 | NP_690607.1 | 2 | 1257 | |||
GenBank mRNA | S72503.1 | - | 2 | 1096 | ||
U07364.1 | - | 2 | 1095 |