Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278717.1 | NP_001265646.1 | 8 | 665 | ||
NM_001278718.1 | NP_001265647.1 | 8 | 559 | |||
NM_005031.4 | NP_005022.2 | 8 | 555 | |||
NM_021902.3 | NP_068702.1 | 8 | 531 | |||
XM_017026874.1 | XP_016882363.1 | 8 | 510 | |||
XM_017026875.1 | XP_016882364.1 | 8 | 725 | |||
XM_017026876.1 | XP_016882365.1 | 8 | 660 | |||
GenBank mRNA | AK289811.1 | - | 8 | 498 | ||
BC032800.2 | - | 7 | 425 | 5179163 | ||
BM691130.1 | - | 8 | 498 | |||
BM856555.1 | - | 8 | 492 | |||
BX115036.1 | - | 8 | 555 |