Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005171.4 | NP_005162.1 | 2, 3 | 409 | ||
XM_011538386.2 | XP_011536688.1 | 2, 3 | 2466 | |||
XM_011538387.2 | XP_011536689.1 | 2, 3 | 139 | |||
XM_017019331.1 | XP_016874820.1 | 2, 3 | 179 | |||
XM_017019332.1 | XP_016874821.1 | 2, 3 | 273 | |||
XM_017019333.1 | XP_016874822.1 | 2, 3 | 159 | |||
XM_017019334.1 | XP_016874823.1 | 2, 3 | 185 | |||
GenBank mRNA | AK313406.1 | - | 2, 3 | 314 | ||
BC029619.1 | - | 2, 3 | 123 | 4102231 | ||
BG502726.1 | - | 2, 3 | 142 | |||
BG545632.1 | - | 2, 3 | 337 | |||
BI712653.1 | - | 2, 3 | 298 | |||
DA468394.1 | - | 2, 3 | 409 |