Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195381.1 | NP_001182310.1 | 6 | 1758 | ||
NM_001195382.1 | NP_001182311.1 | 6 | 1644 | |||
NM_005301.3 | NP_005292.2 | 2 | 1121 | |||
GenBank mRNA | AF089087.1 | - | 1 | 1368 | ||
AK131540.1 | - | 1 | 3304 | |||
AK172786.1 | - | 6 | 1735 | |||
AK290256.1 | - | 6 | 1607 | |||
AK314002.1 | - | 2 | 1121 | |||
BC095500.1 | - | 1 | 790 | |||
BC117453.1 | - | 1 | 1165 | 40126004 | ||
BC117455.1 | - | 1 | 1163 | 40126006 | ||
CR541765.1 | - | 1 | 793 |