Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093770.2 | NP_001087239.2 | 4 | 328 | ||
NM_001164644.1 | NP_001158116.1 | 4 | 298 | |||
NM_001164647.1 | NP_001158119.1 | 3 | 268 | |||
NM_005411.4 | NP_005402.3 | 4 | 339 | |||
XM_005270062.4 | XP_005270119.1 | 3 | 328 | |||
XM_006717953.2 | XP_006718016.1 | 4 | 321 | |||
GenBank mRNA | AK290703.1 | - | 4 | 330 | ||
AK298029.1 | - | 3 | 217 | |||
AK298092.1 | - | 2 | 70 | |||
BC029913.2 | - | 3 | 295 | |||
BC111570.1 | - | 3 | 247 | 40069324 | ||
BC171875.1 | - | 3 | 258 | 9054529 | ||
BI820937.1 | - | 4 | 322 | |||
HQ021433.2 | - | 3 | 268 | |||
HQ021434.1 | - | 3 | 268 | |||
HQ021435.1 | - | 3 | 268 | |||
HQ021436.1 | - | 3 | 268 | |||
HQ021437.1 | - | 4 | 339 | |||
HQ021438.1 | - | 4 | 339 | |||
HQ021439.1 | - | 4 | 339 | |||
HQ021440.1 | - | 4 | 328 | |||
HQ021441.1 | - | 4 | 328 | |||
HQ021442.1 | - | 4 | 328 | |||
JX502764.1 | - | 4 | 339 | |||
JX502765.1 | - | 4 | 328 |