Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006284.3 | NP_006275.1 | 2, 3 | 436 | ||
GenBank mRNA | BC012088.1 | - | 2, 3 | 393 | 4562773 | |
CR541943.1 | - | 2, 3 | 381 | |||
U13991.1 | - | 2, 3 | 398 |