Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPREB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013378.2 | NP_037510.1 | 2 | 191 | ||
GenBank mRNA | AF163825.1 | - | 2 | 153 | ||
AK312098.1 | - | 2 | 146 | |||
AY359000.1 | - | 2 | 191 | |||
BC020666.1 | - | 2 | 216 | 4772032 | ||
BQ899182.1 | - | 2 | 124 | |||
CR456610.1 | - | 2 | 140 |