Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014448.3 | NP_055263.2 | 6, 7 | 1272 | ||
XM_017001049.1 | XP_016856538.1 | 6, 7 | 1873 | |||
XM_017001050.1 | XP_016856539.1 | 6, 7 | 1078 | |||
XM_017001051.1 | XP_016856540.1 | 6, 7 | 1283 | |||
GenBank mRNA | AK095984.1 | - | 6, 7 | 1179 | ||
BC002681.1 | - | 5, 6 | 559 | 3613623 | ||
BC051838.1 | - | 4, 5 | 579 | 6191377 | ||
BT007270.1 | - | 1, 2 | 160 | |||
D89016.1 | - | 3, 4 | 587 | |||
DA539376.1 | - | 5, 6 | 459 |